Ammonium tetrafluoroborate ≥98.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate
M-Clarity™ quality level = MQ200
8.43945.0100
8.43945.1000
8.43945.9025
EM8.43945.0100CS
2527
USD
EM8.43945.0100
EM8.43945.1000
EM8.43945.9025
Ammonium tetrafluoroborate ≥98.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Ammonium tetrafluoroborate
Formula:
H₄BF₄N Density: 1.85 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD06245785 CAS Number: 13826-83-0 UN: 3260 ADR: 8,II |
Specification Test Results
Density | 1.85 g/cm3 (20 °C) |
pH | 3.5 (5 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...