Ammonium tetrafluoroborate ≥98.0% (by acidimetry) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

M-Clarity™ quality level = MQ200

8.43945.0100 8.43945.1000 8.43945.9025
EM8.43945.0100CS 2527 USD
EM8.43945.0100 EM8.43945.1000 EM8.43945.9025
Ammonium tetrafluoroborate ≥98.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Ammonium tetrafluoroborate
Formula: H₄BF₄N
Density: 1.85 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD06245785
CAS Number: 13826-83-0
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Density 1.85 g/cm3 (20 °C)
pH 3.5 (5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR