Polyvinyl alcohol, M.W. ∼ 60000 for synthesis, fully hydrolysed, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Poly(vinyl alcohol), PVOH, PVA, Mowiol®
M-Clarity™ quality level = MQ200
8.43866.0100
8.43866.1000
EM8.43866.0100CS
699
USD
EM8.43866.0100
EM8.43866.1000
Polyvinyl alcohol, M.W. ∼ 60000 for synthesis, fully hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol
Formula:
[-CH₂CH(OH)-]n |
MDL Number:
MFCD00081922 CAS Number: 9002-89-5 |
Specification Test Results
Melting Point | 160 - 240 °C |
Bulk Density | 400 - 600 kg/m3 |
Density | 0.4 - 0.6 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...