D-Glucono-1,5-lactone, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43794.0100
EM8.43794.0100CS 7156 USD
EM8.43794.0100
D-Glucono-1,5-lactone, Sigma-Aldrich®
D-Glucono-1,5-lactone

D-(+)-Glucono-delta-lactone for synthesis

Formula: C₆H₁₀O₆
MDL Number: MFCD00006647
CAS Number: 90-80-2

Order Now

Specification Test Results

Hill Formula C₆H₁₀O₆
Molar Mass 178.14 g/mol
Melting Point 151 - 154 °C (decomposition)
pH value 3.6 (10 g/l, H₂O, 20 °C)
Bulk density 750 kg/m³
Solubility 590 g/l (25 °C) Hydrolysis
Storage class 10 - 13 Other liquids and solids
WGK WGK 1 slightly water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR