Sodium triacetoxyborohydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.43705.0025
EM8.43705.0025CS 2289 USD
EM8.43705.0025 EM8.43705.0100
Sodium triacetoxyborohydride for synthesis, Sigma-Aldrich®
Sodium triacetoxyborohydride
Formula: C₆H₁₀BNaO₆
Melting Pt: 114…118 °C
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 3396
ADR: 4.3 (4.1),II

Order Now

Specification Test Results

Melting Point 110 - 114 °C
Molar Mass 211.94 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR