Phenyltrimethoxysilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Trimethoxy(phenyl)silane

M-Clarity™ quality level = MQ200

8.43658.0050
EM8.43658.0050CS 7795 USD
EM8.43658.0050
Phenyltrimethoxysilane for synthesis, Sigma-Aldrich®
Phenyltrimethoxysilane
Formula: C₉H₁₄O₃Si
Density: 1.064 g/cm³ (20 °C)
MDL Number: MFCD00025689
CAS Number: 2996-92-1
EINECS: 221-066-9

Order Now

Specification Test Results

Molar Mass 198.3 g/mol
Boiling Point 233 °C
Flash Point 83 °C
Density 1.064 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR