(3-Glycidoxypropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
(3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane
M-Clarity™ quality level = MQ200
8.41807.0100
8.41807.0500
EM8.41807.0100CS
1998
USD
EM8.41807.0100
EM8.41807.0500
(3-Glycidoxypropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
(3-Glycidoxypropyl)trimethoxysilane
Formula:
C₉H₂₀O₅Si MW: 236.34 g/mol Boiling Pt: 299 Melting Pt: 0 Density: 1.07 g/cm³ (20 °C) Flash Pt: 122 Storage Temperature: Ambient |
MDL Number:
MFCD00005144 CAS Number: 2530-83-8 EINECS: 219-784-2 |
Specification Test Results
Solubility | (20 °C) immiscible,(decomposition) |
Molar Mass | 236.34 g/mol |
Boiling Point | 290 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 122 °C |
Explosion Limit | 0.43 %(V) |
Density | 1.07 g/cm3 (20 °C) |
pH | 6.8 (100 g/l, H2O) |
Ignition Point | 400 °C |
Viscosity kinematic | 3 mm2/s (25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...