p-Xylylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.41656.0010
EM8.41656.0010CS 76220 USD
EM8.41656.0010
p-Xylylenediamine for synthesis, Sigma-Aldrich®
p-Xylylenediamine
Formula: C₈H₁₂N₂
MDL Number: MFCD00009821
CAS Number: 539-48-0
EINECS: 208-719-3
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Melting Point 60 - 64 °C
Molar Mass 136.19 g/mol
Boiling Point 230 °C (13.33 hPa)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR