3,3-Dimethylbutanone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone
M-Clarity™ quality level = MQ200
8.41645.0100
8.41645.0500
EM8.41645.0100CS
2614
USD
EM8.41645.0100
EM8.41645.0500
3,3-Dimethylbutanone for synthesis, Sigma-Aldrich®
3,3-Dimethylbutanone
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 106 Melting Pt: -53 Density: 0.803 g/cm³ (20 °C) Flash Pt: 5 Storage Temperature: Ambient |
MDL Number:
MFCD00008846 CAS Number: 75-97-8 EINECS: 200-920-4 UN: 1993 ADR: 3 II Merck Index: 12,07594 |
Specification Test Results
Solubility | 19 g/l (25 °C) |
Melting Point | -53 °C |
Molar Mass | 100.16 g/mol |
Boiling Point | 105 - 106 °C (1013 hPa) |
Vapor Pressure | 33 hPa (20 °C) |
Flash Point | 5 °C |
Density | 0.808 g/cm3 (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...