2,3-Dimethylmaleic anhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3,4-Dimethylfuran-2,5-dione

M-Clarity™ quality level = MQ200

8.41502.0005
EM8.41502.0005CS 71065 USD
EM8.41502.0005
2,3-Dimethylmaleic anhydride for synthesis, Sigma-Aldrich®
2,3-Dimethylmaleic anhydride
Formula: C₆H₆O₃
MW: 126.11 g/mol
MDL Number: MFCD00005523
CAS Number: 766-39-2
EINECS: 212-165-8

Order Now

Specification Test Results

Melting Point 96 °C
Molar Mass 126.11 g/mol
Boiling Point 223 °C
Density 1.107 g/cm3 (100 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR