1,10-Phenanthroline, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: o-Phenanthroline

M-Clarity™ quality level = MQ200

8.41491.0005 8.41491.0025
EM8.41491.0005CS 28110 USD
EM8.41491.0005 EM8.41491.0025
1,10-Phenanthroline, anhydrous for synthesis, Sigma-Aldrich®
1,10-Phenanthroline
Formula: C₁₂H₈N₂
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 117…119 °C
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1,II
Merck Index: 12,07356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 2.69 g/l (25 °C)
Melting Point 117 - 119 °C
Molar Mass 180.21 g/mol
Boiling Point 330 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR