Vinyltrimethoxysilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethoxy(vinyl)silane

M-Clarity™ quality level = MQ200

8.41268.0100
EM8.41268.0100CS 2305 USD
EM8.41268.0100
Vinyltrimethoxysilane for synthesis, Sigma-Aldrich®
Vinyltrimethoxysilane
Formula: C₅H₁₂O₃Si
MW: 148.23 g/mol
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: 0 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008605
CAS Number: 2768-02-7
EINECS: 220-449-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Molar Mass 148.23 g/mol
Boiling Point 122 - 124 °C
Vapor Pressure 88 hPa (55 °C)
Flash Point 22 °C
Explosion Limit 1.1 %(V)
Density 0.97 g/cm3 (20 °C)
Ignition Point 235 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR