4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.40130.0025
8.40130.0100
EM8.40130.0025CS
6728
USD
EM8.40130.0025
EM8.40130.0100
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Formula:
C₉H₁₈NO₂ |
MDL Number:
MFCD00006478 CAS Number: 2226-96-2 EINECS: 218-760-9 |
Specification Test Results
Solubility | 1670 g/l (20 °C) |
Melting Point | 67 - 71 °C |
Molar Mass | 172.25 g/mol |
Boiling Point | (1013 hPa) not applicable,(decomposition) |
Vapor Pressure |
|
Flash Point | 146 °C DIN 51758 |
pH | 8.2 (20 g/l, H2O, 20 °C) |
Ignition Point | 260 °C DIN 51794 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...