Platinum(II) chloride Pt 73% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Platinum dichloride

M-Clarity™ quality level = MQ200

8.24566.0500
EM8.24566.0500CS 703153 USD
EM8.24566.0500
Platinum(II) chloride Pt 73% for synthesis, Sigma-Aldrich®
Platinum (II) chloride
Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 °C
Density: 6.05 g/cm³ (20 °C)
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 1759
ADR: 8,III
Merck Index: 12,07687

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 581 °C decomposes
Molar Mass 265.99 g/mol
Density 6.1 g/cm3
pH 5 - 6 (H2O) suspension

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR