Triethanolamine (trolamine), EMPLURA®, Supelco®

Supplier: MilliporeSigma

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

M-Clarity™ quality level = MQ200

8.22341.9026
EM8.22341.9026EA 658 USD
EM8.22341.9026
Triethanolamine (trolamine), EMPLURA®, Supelco®
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
Boiling Pt: 335.4 °C (1013 hPa)
Melting Pt: 21.6 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR