Hydroxylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

M-Clarity™ quality level = MQ200

8.22334.0250
EM8.22334.0250CS 1140 USD
EM8.22334.0250 EM8.22334.1000
Hydroxylammonium chloride for synthesis, Sigma-Aldrich®
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
MDL Number: MFCD00051089
CAS Number: 5470-11-1
UN: 2923
ADR: 8 (6.1),III

Order Now

Specification Test Results

Grade Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Synonyms Hydroxylammonium Chloride
Chemical Formula NH2OH·HCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR