Hydroquinone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.22333.0250 8.22333.1000 8.22333.2501
EM8.22333.0250CS 863 USD
EM8.22333.0250 EM8.22333.1000 EM8.22333.2501
Hydroquinone for synthesis, Sigma-Aldrich®
Hydroquinone
Formula: C₆H₄(OH)₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 172 °C
Density: 1.332 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9,III
Merck Index: 13,04833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203cfb319003b2b4489cbe3ab15876fc37f27cb8&vid=6d76260c677491e292c03637f8a08e9194282d48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 70 g/l (25 °C)
Melting Point 172 °C (decomposition)
Molar Mass 110.11 g/mol
Bulk Density 600 kg/m3
Boiling Point 287 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (25 °C)
Flash Point 165 °C
Density 1.358 g/cm3 (20 °C)
pH (70 g/l, H2O, 20 °C)
Ignition Point 515 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR