(±)-2-Butanol, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.22263.2500
EM8.22263.2500CS 490 USD
EM8.22263.2500
(±)-2-Butanol, EMPLURA®, Supelco®
(±)-2-Butanol
Formula: H₃CCH₂CH(OH)CH₃
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.8534 g/cm³ (20 °C)
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,III
Merck Index: 13,01540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR