Tetraethylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Tetrylammonium hydroxide

M-Clarity™ quality level = MQ200

8.22149.9025
EM8.22149.9025EA 1658 USD
EM8.22149.9025
Tetraethylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®
Tetraethylammonium hydroxide
Formula: C₈H₂₁NO
MW: 147.26
Density: 0.865
MDL Number: MFCD00009024
CAS Number: 77-98-5
UN: 3267
ADR: 8 II
Merck Index: 12,09342

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR