D-(+)-Sucrose octaacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.22144.0250
EM8.22144.0250CS 1740 USD
EM8.22144.0250
D-(+)-Sucrose octaacetate for synthesis, Sigma-Aldrich®
D(+)-Sucrose octaacetate
Formula: C₂₈H₃₈O₁₉
MDL Number: MFCD00006623
CAS Number: 126-14-7

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 82 - 87 °C
Molar Mass 678.59 g/mol
Boiling Point 250 °C (1.3 hPa)
Synonyms Octaacetylsucrose
Hill Formula C28H38O19


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR