D-Gluconic acid 50% in aqueous solution, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Gluconic acid, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid

M-Clarity™ quality level = MQ200

8.22057.9025
EM8.22057.9025EA 713 USD
EM8.22057.9025
D-Gluconic acid 50% in aqueous solution, Sigma-Aldrich®
D-Gluconic acid
Formula: C₆H₁₂O₇
MW: 196.16 g/mol
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00004240
CAS Number: 526-95-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR