Sodium dodecyl sulfate (SDS) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
M-Clarity™ quality level = MQ200
8.22050.1000
8.22050.0100
8.22050.2500
8.22050.9020
EM8.22050.1000EA
397
USD
EM8.22050.1000
EM8.22050.0100
EM8.22050.2500
EM8.22050.9020
Sodium dodecyl sulfate (SDS) for synthesis, Sigma-Aldrich®
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 204…207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,08636 |
|
Specification Test Results
Solubility | 150 g/l (20 °C) |
Melting Point | 204.06 - 207.06 °C |
Molar Mass | 288.4 g/mol |
Bulk Density | 490 - 560 kg/m3 |
Vapor Pressure | (20 °C) |
Flash Point |
|
Density | 1.1 g/cm3 (20 °C) |
pH | 6 - 9 (10 g/l, H2O, 20 °C) |
Hazard Class | 4.1 |
Hazard Class Description | Flammable Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...