Pyrene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21051.0250
EM8.21051.0005CS 37527 USD
EM8.21051.0005 EM8.21051.0050 EM8.21051.0250
Pyrene for synthesis, Sigma-Aldrich®
Pyrene
Formula: C₁₆H₁₀
MDL Number: MFCD00004136
CAS Number: 129-00-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b03eb6b87b60315a282988993f066fd2d3af129&vid=d3f1ffa103d650d2e9f49139db04842dfa6ca0ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.135 mg/l (25 °C)
Melting Point 148 - 150 °C
Molar Mass 202.26 g/mol
Bulk Density 650 kg/m3
Boiling Point 393 °C (1013 hPa)
Flash Point 200 - 220 °C
Density 1.21 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR