Polyethylene glycol 2,000 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.21037.1000 8.21037.5000
EM8.21037.1000CS 481 USD
EM8.21037.1000 EM8.21037.5000
Polyethylene glycol 2,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 2,000
Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Specification Test Results

Solubility 600 g/l (20 °C)
Melting Point 49 - 52 °C
Molar Mass 1800.00 g/mol
Bulk Density 400 - 500 kg/m3
Flash Point 250 °C
Density 1.21 g/cm3 (20 °C)
pH 4 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 420 °C
Viscosity kinematic 46 - 55 mm2/s (20 °C) (50% solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR