Perylene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20969.0001 8.20969.0005
EM8.20969.0001CS 71578 USD
EM8.20969.0001 EM8.20969.0005
Perylene for synthesis, Sigma-Aldrich®
Perylene
Formula: C₂₀H₁₂
MW: 252.32 g/mol
Boiling Pt: 443
MDL Number: MFCD00004142
CAS Number: 198-55-0
Merck Index: 12,07325

Order Now

Specification Test Results

Solubility  - 0.0001 g/l (25 °C)
Melting Point 273 - 278 °C
Molar Mass 252.31 g/mol
Vapor Pressure  - 0.001 hPa (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR