Pentafluoropropionic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid

M-Clarity™ quality level = MQ200

8.20956.0010 8.20956.0050
EM8.20956.0010CS 86414 USD
EM8.20956.0010 EM8.20956.0050
Pentafluoropropionic acid for synthesis, Sigma-Aldrich®
Pentafluoropropionic acid
Formula: C₃HF₅O₂
MDL Number: MFCD00004170
CAS Number: 422-64-0
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 164.03 g/mol
Boiling Point 96 - 97 °C (984 hPa)
Vapor Pressure 52 hPa (25 °C)
Refractive Index 1.2838 (25 °C, 589 nm)
Density 1.57 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR