(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20819.9025
EM8.20819.9025EA 468 USD
EM8.20819.9025
(±)-2-Methyl-2,4-pentanediol, Sigma-Aldrich®
(±)-2-Methyl-2,4-pentanediol
Formula: C₆H₁₄O₂
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.923 g/cm³ (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00004547
CAS Number: 107-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR