Indene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1H-Indene

M-Clarity™ quality level = MQ200

8.20701.0250 8.20701.1000
EM8.20701.0250CS 833 USD
EM8.20701.0250 EM8.20701.1000
Indene for synthesis, Sigma-Aldrich®
Indene
Formula: C₉H₈
MDL Number: MFCD00003777
CAS Number: 95-13-6
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -2 °C
Molar Mass 116.16 g/mol
Boiling Point 181 - 183 °C
Vapor Pressure 1.7 hPa (20 °C)
Flash Point 58 °C
Refractive Index 1.5762 (20 °C, 589 nm)
Density 0.99 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR