Diphenylcyclopropenone, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

M-Clarity™ quality level = MQ200

8.20533.8500
EM8.20533.8500EA 44 USD
EM8.20533.8500
Diphenylcyclopropenone, Sigma-Aldrich®
Diphenylcyclopropenone
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 407
Melting Pt: 112…113 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00001311
CAS Number: 886-38-4
Merck Index: 12,03366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR