Diphenylcyclopropenone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
M-Clarity™ quality level = MQ200
8.20533.0001
8.20533.0005
EM8.20533.0001CS
8916
USD
EM8.20533.0001
EM8.20533.0005
Diphenylcyclopropenone for synthesis, Sigma-Aldrich®
Diphenylcyclopropenone
Formula:
C₁₅H₁₀O MW: 206.24 g/mol Boiling Pt: 407 |
MDL Number:
MFCD00001311 CAS Number: 886-38-4 Merck Index: 12,03366 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 118 - 122 °C |
Molar Mass | 206.24 g/mol |
Hill Formula | C15H10O |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...