1-Chloronaphthalene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20315.0100 8.20315.1000
EM8.20315.0100CS 837 USD
EM8.20315.0100 EM8.20315.1000
1-Chloronaphthalene for synthesis, Sigma-Aldrich®
1-Chloronaphthalene
Formula: C₁₀H₇Cl
MDL Number: MFCD00003874
CAS Number: 90-13-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.009 g/l (20 °C)
Melting Point -6 °C
Molar Mass 162.61 g/mol
Boiling Point 259 °C (1013 hPa)
Vapor Pressure 0.05 hPa (20 °C)
Flash Point 115 °C
Density 1.19 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) (undiluted)
Ignition Point 500 °C
Synonyms α-Naphthyl chloride
Hill Formula C10H7Cl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR