1-Chloro-2-methylpropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20314.0050 8.20314.0250
EM8.20314.0050CS 4114 USD
EM8.20314.0050 EM8.20314.0250
1-Chloro-2-methylpropane for synthesis, Sigma-Aldrich®
1-Chloro-2-methylpropane
Formula: C₄H₉Cl
MDL Number: MFCD00000954
CAS Number: 513-36-0
UN: 1127
ADR: 3,II

Order Now

Specification Test Results

Solubility 28 mg/l (20 °C)
Melting Point -131 °C
Molar Mass 92.56 g/mol
Boiling Point 69 °C (1013 hPa)
Vapor Pressure 161 hPa (20 °C)
Flash Point -17 °C
Refractive Index 1.3975 (20 °C, 589 nm)
Explosion Limit 2.0 - 8.8 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 395 °C
Synonyms Isobutyl chloride
Hill Formula C4H9Cl
Chemical Formula (CH3)2CHCH2Cl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR