2-Chloroethylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

M-Clarity™ quality level = MQ200

8.20265.0100 8.20265.0250
EM8.20265.0100CS 1322 USD
EM8.20265.0100 EM8.20265.0250
2-Chloroethylammonium chloride for synthesis, Sigma-Aldrich®
2-Chloroethylammonium chloride
Formula: C₂H₇Cl₂N
MW: 115.99 g/mol
MDL Number: MFCD00012887
CAS Number: 870-24-6
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Solubility 5666 g/l (20 °C)
Melting Point 143 - 146 °C
Molar Mass 115.99 g/mol
Bulk Density 750 - 850 kg/m3
pH 2 - 3 (5666 g/l, H2O, 20 °C)
Synonyms 2-Chloroethylamine hydrochloride, ß-Aminoethyl chloride hydrochloride
Hill Formula C2H7Cl2N
Chemical Formula ClCH2CH2NH2 * HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR