Dibutyl disulfide, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20242.0010
EM8.20242.0010EA 39 USD
EM8.20242.0010
Dibutyl disulfide, Sigma-Aldrich®
Dibutyl disulphide
Formula: C₈H₁₈S₂
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: –94 °C
Density: 0.9371 g/cm³ (20 °C)
MDL Number: MFCD00009467
CAS Number: 629-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR