2-Bromethylamine hydrobromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

M-Clarity™ quality level = MQ200

8.20176.0100 8.20176.0500 8.20176.5000
EM8.20176.0100CS 2356 USD
EM8.20176.0100 EM8.20176.0500 EM8.20176.5000
2-Bromethylamine hydrobromide for synthesis, Sigma-Aldrich®
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 171 - 176 °C
Molar Mass 204.89 g/mol
pH 4.7 (25 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR