Isoamyl alcohol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18969.1000
EM8.18969.1000CS 472 USD
EM8.18969.1000
Isoamyl alcohol for synthesis, Sigma-Aldrich®
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MDL Number: MFCD00002934
CAS Number: 123-51-3
UN: 1105
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 25 g/l (20 °C)
Melting Point -117 °C
Molar Mass 88.15 g/mol
Boiling Point 131 °C (1013 hPa)
Vapor Pressure 3 hPa (20 °C)
Flash Point 45 °C
Explosion Limit 1.2 - 8 %(V)
Density 0.81 g/cm3 (20 °C)
pH 7 (25 g/l, H2O, 20 °C)
Ignition Point 335 °C
Saturation concentration 11 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR