Polyethylene glycol 35,000 stabilized for synthesis pH 5 - 7 (100 g/l, H₂O, 20 °C), Sigma-Aldrich®

Supplier: MilliporeSigma


Stabilization: Stabilised with 2-tert-butyl-4-methoxyphenol
M-Clarity™ quality level = MQ200

8.18892.1000 8.18892.5000
EM8.18892.1000EA 87 USD
EM8.18892.1000 EM8.18892.5000
Polyethylene glycol 35,000 stabilized for synthesis pH 5 - 7 (100 g/l, H₂O, 20 °C), Sigma-Aldrich®
Polyethylene glycol 35,000
Formula: H(OCH₂CH₂)nOH
Melting Pt: 60…65 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Hydroxyl value 3-4
Melting range (lower value) ≥ 62 °C
Melting range (upper value) ≤ 67 °C
Average molecular mass 28000 - 38000
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR