Hydrobromic acid 33% in acetic acid, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrogen bromide

M-Clarity™ quality level = MQ200

8.18856.1000
EM8.18856.1000EA 206 USD
EM8.18856.1000
Hydrobromic acid 33% in acetic acid, Sigma-Aldrich®
Hydrogen bromide
Formula: HBr
MW: 80.91 g/mol
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 2920
ADR: 8 (3),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR