Hydrobromic acid 33% in acetic acid, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hydrogen bromide

M-Clarity™ quality level = MQ200

8.18856.0250
EM8.18856.0250CS 684 USD
EM8.18856.0250
Hydrobromic acid 33% in acetic acid, Sigma-Aldrich®
Hydrogen bromide
Formula: HBr
MW: 80.91 g/mol
MDL Number: MFCD00011323
CAS Number: 10035-10-6

Order Now

Specification Test Results

Bulk Density (20 °C) soluble
Boiling Point  - 200 °C (1013 hPa)
Vapor Pressure 360 hPa (20 °C)
Density 1.38 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR