2-Decanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Decan-2-one

M-Clarity™ quality level = MQ200

8.18795.0025
EM8.18795.0025CS 8214 USD
EM8.18795.0025
2-Decanone for synthesis, Sigma-Aldrich®
2-Decanone
Formula: C₁₀H₂₀O
MDL Number: MFCD00009571
CAS Number: 693-54-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 3 °C
Molar Mass 156.26 g/mol
Boiling Point 209 - 212 °C
Flash Point 86 °C
Refractive Index 1.4253 (20 °C, 589 nm)
Density 0.82 g/cm3 (20 °C)
Synonyms Methyl octyl ketone
Hill Formula C10H20O
Chemical Formula CH3(CH2)7COCH3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR