Tetrabutylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18759.0050
EM8.18759.0050EA 46 USD
EM8.18759.0050
Tetrabutylammonium hydroxide 20% in aqueous solution, Sigma-Aldrich®
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR