Vanillin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

M-Clarity™ quality level = MQ200

8.18718.0500
EM8.18718.0500CS 826 USD
EM8.18718.0500
Vanillin for synthesis, Sigma-Aldrich®
Vanillin
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285
Melting Pt: 82
Flash Pt: 153
MDL Number: MFCD00006942
CAS Number: 121-33-5
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 10 g/l (25 °C)
Melting Point 82 °C
Molar Mass 152.14 g/mol
Bulk Density 600 kg/m3
Boiling Point 154 °C (13 hPa)
Vapor Pressure 0.0029 hPa (25 °C)
Flash Point 153 °C
Density 1.06 g/cm3 (20 °C)
pH 4.3 (10 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR