Formaldehyde 37% in aqueous solution stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: Stabilized with methanol 10%
M-Clarity™ quality level = MQ200

8.18708.5000
EM8.18708.5000CS 525 USD
EM8.18708.5000
Formaldehyde 37% in aqueous solution stabilized for synthesis, Sigma-Aldrich®
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

Density 1.017 g/cm3 (20 °C)
pH 0.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR