n-Undecane, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Undecane

M-Clarity™ quality level = MQ200

8.18629.0050
EM8.18629.0050EA 56 USD
EM8.18629.0050
n-Undecane, Sigma-Aldrich®
n-Undecane
Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –26.7 °C
Density: 0.7416 g/cm³ (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00008959
CAS Number: 1120-21-4
UN: 2330
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR