Thiourea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18591.0500
EM8.18591.0500CS 1267 USD
EM8.18591.0500
Thiourea for synthesis, Sigma-Aldrich®
Thiourea
Formula: NH₂CSNH₂
MDL Number: MFCD00008067
CAS Number: 62-56-6
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 137 g/l (20 °C)
Melting Point 169 - 173 °C
Molar Mass 76.12 g/mol
Bulk Density 640 kg/m3
Vapor Pressure (20 °C)
Density 1.405 g/cm3 (20 °C)
pH 6 - 8 (50 g/l, H2O, 20 °C)
Ignition Point 440 °C Dust
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR