Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, Nerolidol, 3,7,11-Trimethyldodeca-1,6,10-trien-3-ol

M-Clarity™ quality level = MQ200

8.18553.0100
EM8.18553.0100CS 1998 USD
EM8.18553.0100
Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®
Nerolidol (cis and trans mixture)
Formula: C₁₅H₂₆O
MW: 222.37 g/mol
MDL Number: MFCD00008911
CAS Number: 7212-44-4
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.014 g/l (20 °C)
Melting Point -75 °C
Molar Mass 222.37 g/mol
Boiling Point 263 - 266 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 96 °C
Refractive Index 1.4893 (20 °C, 589 nm)
Explosion Limit 0.7 - 4.5 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 230 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR