(3-Aminopropyl)trimethoxysilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane

M-Clarity™ quality level = MQ200

8.18550.0250
EM8.18550.0250CS 2261 USD
EM8.18550.0250
(3-Aminopropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
(3-Aminopropyl)trimethoxysilane
Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
MDL Number: MFCD00008206
CAS Number: 13822-56-5

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 179.29 g/mol
Boiling Point 194 °C (1013 hPa)
Flash Point 90 °C
Density 1.02 g/cm3 (20 °C)
pH 9 (20 g/l, H2O, 20 °C)
Ignition Point 295 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR