(±)-3-Amino-1,2-propanediol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18522.0025
EM8.18522.0025CS 7077 USD
EM8.18522.0025
(±)-3-Amino-1,2-propanediol for synthesis, Sigma-Aldrich®
(±)-3-Amino-1,2-propanediol
Formula: C₃H₉NO₂
MDL Number: MFCD00008140
CAS Number: 616-30-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 91.11 g/mol
Boiling Point 140 - 141 °C (12 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 186 °C
Refractive Index 1.492 (20 °C, 589 nm)
Density 1.19 g/cm3 (20 °C)
pH 11 (100 g/l, H2O, 20 °C)
Synonyms 1-Amino-2,3-propanediol
Hill Formula C3H9NO2
Chemical Formula NH2CH2CH(OH)CH2OH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR