5-Fluorouracil for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

M-Clarity™ quality level = MQ200

8.18505.0005
EM8.18505.0005CS 79731 USD
EM8.18505.0005
5-Fluorouracil for synthesis, Sigma-Aldrich®
5-Fluorouracil
Formula: C₄H₃FN₂O₂
MDL Number: MFCD00006018
CAS Number: 51-21-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 12 g/l (20 °C)
Melting Point 282 - 283 °C (decomposition)
Molar Mass 130.08 g/mol
Boiling Point 190 - 200 °C (0.13 hPa)
pH 4.3 - 5.3 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR