(4-Carboxybutyl)triphenylphosphonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: (4-Carboxybut-1-yl)(triphenyl)phosphonium bromide

M-Clarity™ quality level = MQ200

8.18330.0050
EM8.18330.0050CS 2562 USD
EM8.18330.0050
(4-Carboxybutyl)triphenylphosphonium bromide for synthesis, Sigma-Aldrich®
(4-Carboxybutyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrO₂P
MW: 443.32 g/mol
MDL Number: MFCD00011906
CAS Number: 17814-85-6
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 204 - 208 °C
Molar Mass 443.31 g/mol
Bulk Density 360 - 400 kg/m3
Flash Point 195 °C
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR